Psoralen 97%
Supplier: ALFA AESAR
|
Warning
|
J60042.MC
J60042.ME
J60042.MCEA
269
EUR
J60042.MC
J60042.ME
Psoralen 97%
Psoralen
Purity: 97%
CAS No.: 66-97-7
Molecular Formula: C11H6O3
Formula Weight: 186.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
Melting Point: 163-164°
Beilstein Reference: 152784
MDL No.: MFCD00010520
Applications: Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure
Formula:
C₁₁H₆O₃ Melting Pt: 163…164 °C |
MDL Number:
MFCD00010520 CAS Number: 66-97-7 EINECS: 200-639-7 Merck Index: 14,7928 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...