1-(1-Naphthyl)piperazine hydrochloride
Supplier: ALFA AESAR
|
Warning
|
J62097.03
J62097.06
J62097.03EA
224
EUR
J62097.03
J62097.06
1-(1-Naphthyl)piperazine hydrochloride
1-(1-Naphthyl)piperazine hydrochloride
CAS No.: 104113-71-5
Molecular Formula: C14H16N2.HCl
Formula Weight: 248.75
Physical Form: Solid
Appearance: Purple
MDL No.: MFCD00055161
Applications: 5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist
Formula:
C₁₄H₁₆N₂·HCl MW: 248.75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055161 CAS Number: 104113-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...