TRIS-acetate-SDS running buffer (10X), pH 8,3
J62560.AP
J62560.K2
J62560.APEA
64.4
EUR
J62560.AP
J62560.K2
TRIS-acetate-SDS running buffer (10X), pH 8,3
Buffers
Electrophoresis Buffers
Running Buffers
Physical Form: Liquid
Applications: Running buffer for TRIS-acetate gels
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...