BSA blocking buffer, 5% in TBS
Supplier: ALFA AESAR
J62637.AK
J62637.AP
J62637.AKEA
0
EUR
J62637.AK
J62637.AP
BSA blocking buffer, 5% in TBS
Buffers
Electrophoresis Buffers
Blocking Buffers
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...