4-Chloro-3-nitrocoumarin 98%
Supplier: ALFA AESAR
|
Warning
|
L07908.03
L07908.06
L07908.03EA
0
EUR
L07908.03
L07908.06
4-Chloro-3-nitrocoumarin 98%
4-Chloro-3-nitrocoumarin
Purity: 98%
CAS No.: 38464-20-9
Molecular Formula: C9H4ClNO4
Formula Weight: 225.59
Melting Point: ca 162° dec.
Beilstein Reference: 883649
MDL No.: MFCD00051670
Formula:
C₉H₄ClNO₄ Melting Pt: ∼ 162 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051670 CAS Number: 38464-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...