Methyl hydrogen azelate 96%

L08116.06 L08116.14
L08116.06EA 72.2 EUR
L08116.06 L08116.14
Methyl hydrogen azelate 96%
Methyl hydrogen azelate

Purity: 96% CAS No.: 2104-19-0 Molecular Formula: C10H18O4 Formula Weight: 202.25 Flash Point: 110°(230°F) Refractive Index: 1.4470 Melting Point: 22-24° Boiling Point: 146-147°/1mm Beilstein Reference: 1775786 MDL No.: MFCD00004431

Formula: C₁₀H₁₈O₄
Boiling Pt: 146…147 °C (1 mmHg)
Melting Pt: 22…24 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004431
CAS Number: 2104-19-0
EINECS: 218-275-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR