Hexyl acrylate 95% stabilised
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ)
L09905.14
L09905.22
L09905.14EA
33.3
EUR
L09905.14
L09905.22
Hexyl acrylate 95% stabilised
Hexyl acrylate
Stab. with hydroquinone
Purity: 95%
CAS No.: 2499-95-8
Molecular Formula: C9H16O2
Formula Weight: 156.22
Flash Point: 68°(154°F)
Refractive Index: 1.4280
Melting Point: -45°
Boiling Point: 88-89°/24mm
Beilstein Reference: 1757327
UN No.: UN3082
MDL No.: MFCD00048881
Formula:
C₉H₁₆O₂ MW: 156.23 g/mol Boiling Pt: 88…89 °C (24 mmHg) Melting Pt: –45 °C Density: 0.890 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048881 CAS Number: 2499-95-8 EINECS: 219-698-5 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...