Dowex® 50WX4 (H) 200-400 mesh
L13918.18
L13918.30
L13918.0B
L13918.18EA
60.8
EUR
L13918.18
L13918.30
L13918.0B
Dowex® 50WX4 (H) 200-400 mesh
Dowex® 50WX4 (H)
CAS No.: 11113-61-4
MDL No.: MFCD00132725
Storage Temperature:
Ambient |
MDL Number:
MFCD00132725 CAS Number: 11113-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...