4,4'-Bipiperidine dihydrochloride 99%

Warning

L15696.06 L15696.14
L15696.06EA 51.5 EUR
L15696.06 L15696.14
4,4'-Bipiperidine dihydrochloride 99%
4,4'-Bipiperidine dihydrochloride

Purity: 99% CAS No.: 78619-84-8 Molecular Formula: C10H20N2.2HCl Formula Weight: 241.21 Melting Point: 300° Beilstein Reference: 3687878 MDL No.: MFCD00005998

Formula: C₁₀H₂₀N₂·2HCl
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005998
CAS Number: 78619-84-8
EINECS: 278-958-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR