Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥98%

L16891.03 L16891.06
L16891.03EA 83 EUR
L16891.03 L16891.06
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥98%
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether

Purity: 98+% CAS No.: 28523-86-6 Molecular Formula: (CF3)2CHOCH2F Formula Weight: 200.06 Boiling Point: 58° Beilstein Reference: 2041023 MDL No.: MFCD00153189

Formula: C₄H₃F₇O
Boiling Pt: 58 °C
Density: 1,505
Storage Temperature: Ambient
MDL Number: MFCD00153189
CAS Number: 28523-86-6
Merck Index: 14,8475

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR