Fmoc-2-amino-4-chlorobenzoic acid 95%
|
Warning
|
L19809.03
L19809.06
L19809.03EA
71.4
EUR
L19809.03
L19809.06
Fmoc-2-amino-4-chlorobenzoic acid 95%
Fmoc-2-amino-4-chlorobenzoic acid
Purity: 95%
CAS No.: 332121-92-3
Molecular Formula: C22H16ClNO4
Formula Weight: 393.83
Melting Point: 218-220° dec.
MDL No.: MFCD02682157
Formula:
C₂₂H₁₆ClNO₄ Melting Pt: 218…220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02682157 CAS Number: 332121-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...