Silicone antifoam emulsion AF, food grade
Warning
A1303-200LTBL A1303-4LTPL
SPCMA1303-200LTBLEA 0 EUR
SPCMA1303-200LTBL SPCMA1303-4LTPL
Silicone antifoam emulsion AF, food grade
Silicone antifoam
Persistent foam-control agent. Also known as polydimethylsiloxane.
Part of a group of polymeric organosilicon compounds known as silicones. This chemical additive reduces and hinders the formation of foam in industrial process liquids.
Antifoam AF is a food grade emulsion.