Dibutyl phosphate 97%

Danger

406645000 406640025 406642500
ACRO406645000EA 94.9 EUR
ACRO406645000 ACRO406640025 ACRO406642500
Dibutyl phosphate 97%
Dibutyl phosphate

Appearance: Clear liquid

Formula: C₈H₁₉O₄P
MW: 210.21 g/mol
Boiling Pt: 250 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.058 g/cm³
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00015245
CAS Number: 107-66-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4290 (20°C, 589 nm)
Water ≤0.3 %
Color scale ≤100 APHA
Titration with NaOH ≥96.0 %
Impurity ≤3.0 % Tributyl phosphate
Acid value 259 to 267 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR