Oxamic hydrazide 99%

129640250 129641000
ACRO129640250EA 54.4 EUR
ACRO129640250 ACRO129641000
Oxamic hydrazide 99%
Oxamic hydrazide

CAS No.: 515-96-8

Formula: C₂H₅N₃O₂
Melting Pt: 216…218 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008004
CAS Number: 515-96-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR