Pivalonitrile 98%

Supplier: ACROS ORGANICS
Danger

140030250 140031000
ACRO140030250EA 73.2 EUR
ACRO140030250 ACRO140031000
Pivalonitrile 98%
Pivalonitrile

Appearance: Clear colorless to light yellow Liquid

Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 105…106 °C (760 mmHg)
Melting Pt: 15…16 °C
Density: 0.75 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001847
CAS Number: 630-18-2
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR