Dowex® 1X2 (Cl) 100-200 mesh resin_ion-exchange

202961000 202965000
ACRO202961000EA 107 EUR
ACRO202961000 ACRO202965000
Dowex® 1X2 (Cl) 100-200 mesh resin_ion-exchange
Dowex® 1X2 (Cl)

CAS No.: 9085-42-1

Storage Temperature: Ambient
MDL Number: MFCD00132715
CAS Number: 9085-42-1

Order Now

Specification Test Results

Appearance Beads
Particle size ≤10 % (> 300 µm)
Wet volume capacity ≥0.6 meq/ml (Cl)
Water retention capacity 65 to 75 % (Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR