Chemical, flame and electric arc protective overalls, Tychem® 6000 FR ThermoPro, model TP198T OR
Supplier: DuPont
TP198TORSM0002 TP198TORMD0002 TP198TORLG0002 TP198TOR4X0002 TP198TORXL0002 TP198TOR2X0002 TP198TOR3X0002
113-8703EA 1590 EUR
113-8703 113-8702 113-8701 113-8700 113-8704 113-8698 113-8699
Chemical, flame and electric arc protective overalls, Tychem® 6000 FR ThermoPro, model TP198T OR
Disposable Garments
This Tychem® garment provides triple hazard protection from liquid-chemical splash, flash fire and electric arc in a single layer. It combines the trusted chemical protection of Tychem® and thermal/arc protection of Nomex® to help provide industrial workers and HazMat responders with permeation protection against a broad range of toxic industrial chemicals and to provide escape time from flash fires.
  • Respirator-fit hood with drawstrings, elasticated wrists and hemmed open ankles
  • Long zip extends to chin for complete coverage of neck area
  • Double storm flap, zip and hook and loop closure system for higher protection
  • Sewn with Dupont™ Nomex® thread
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Bright orange colour for high visbility
  • Anti-static treatment (EN 1149-5) on inside
Typical applications include use in the oil and gas industry, by hazmat teams in fire departments and industrial fire brigades, industrial chemical processing plants, and laboratories.
Weight: 280 g/m²
Certifications: Type 3/4/6, EN 1149-5, EN ISO 11612 (heat and flame), EN ISO 14116 (limited flame spread), IEC 61482-2 (electric arc), EN ISO 11611 (welding and allied processes); Cat. III; NFPA 1992, NFPA 2112, Cat 2; NFPA 70E