(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%

Warning

375100010 375100050
ACRO375100010EA 92.5 EUR
ACRO375100010 ACRO375100050
(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%
(S)-(-)-2-(Boc-amino)-1,4-butanediol
Formula: C₉H₁₉NO₄
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD03093946
CAS Number: 128427-10-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 65 to 69 °C
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20 °C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR