4-Chlorophenylmagnesium bromide 1 M in diethyl ether, AcroSeal®

Danger

377401000 377408000
377401000.EA 82.7 EUR
377401000. 377408000.
4-Chlorophenylmagnesium bromide 1 M in diethyl ether, AcroSeal®
4-Chlorophenylmagnesium bromide
Formula: C₆H₄BrClMg
MW: 215.76 g/mol
Density: 0.85 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009926
CAS Number: 873-77-8
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Light yellow to grey to brown-black Solution (May develop some turbidity or precipitate)
Titration Grignard 0.9 to 1.2 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR