4-(Dibutylamino)benzaldehyde 98%

Warning

439770010 439770050
ACRO439770010EA 52.9 EUR
ACRO439770010 ACRO439770050
4-(Dibutylamino)benzaldehyde 98%
4-(Dibutylamino)benzaldehyde
Formula: C₁₅H₂₃NO
MW: 233.355 g/mol
Boiling Pt: 148 °C (1Torr)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075134
CAS Number: 90134-10-4

Order Now

Specification Test Results

Appearance Yellow to brown Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR