N-tert-Butylcyclohexylamine 97%
|
Danger
|
439830010
439830050
ACRO439830010EA
67.4
EUR
ACRO439830010
ACRO439830050
N-tert-Butylcyclohexylamine 97%
N-tert-Butylcyclohexylamine
Formula:
C₁₀H₂₁N Boiling Pt: 172…174 °C (760 mmHg) Density: 0,834 g/cm³ Flash Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075598 CAS Number: 51609-06-4 UN: 2734 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...