(±)-Methyl-2-methylbutyrate 98%

Supplier: ACROS ORGANICS
Danger

450230010 450230050 450231000
ACRO450230010EA 93.5 EUR
ACRO450230010 ACRO450230050 ACRO450231000
(±)-Methyl-2-methylbutyrate 98%
(±)-Methyl 2-methylbutyrate
Formula: C₆H₁₂O₂
Boiling Pt: 115 °C (760 mmHg)
Density: 0,885 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00009335
CAS Number: 868-57-5
UN: 3273
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3930 to 1.3960 (20°C, 589 nm)
GC ≥97.5 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR