3-Oxetanone 95%

Warning

451200010 451200050
ACRO451200010EA 99.2 EUR
ACRO451200010 ACRO451200050
3-Oxetanone 95%
3-Oxetanone
Formula: C₃H₄O₂
Boiling Pt: 140 °C
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD09263255
CAS Number: 6704-31-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR