Anti-CD70 Mouse Monoclonal Antibody (CF594) [clone: BU69]

Supplier: Biotium
BNC940187-500 BNC940187-100
BNC940187-500EA 560 EUR
BNC940187-500 BNC940187-100
Anti-CD70 Mouse Monoclonal Antibody (CF594) [clone: BU69]
Antibodies
This antibody recognizes a protein of 30 kDa, identified as CD70. It is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T- and B-lymphocytes. It induces proliferation of co-stimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis. This MAb blocks the interaction between CD27 and CD70, and has been shown toinhibit T cell proliferation induced by dendritic cells.

Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: BU69
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR