Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: HRB2/273]

Supplier: Biotium
BNC400273-500 BNC400273-100
BNC400273-500EA 560 EUR
BNC400273-500 BNC400273-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: HRB2/273]
Antibodies
This MAb is specific to c-erbB-2/HER-2 and shows minimal cross-reaction with other members of the family. C-erbB-2/HER-2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/273
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR