Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: BH1]

Supplier: Biotium
BNCB0320-500 BNCB0320-100
BNCB0320-500EA 560 EUR
BNCB0320-500 BNCB0320-100
Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: BH1]
Antibodies
CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: BH1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR