Anti-CD38 Mouse Monoclonal Antibody (CF640R) [clone: AT1]

Supplier: Biotium
BNC400430-500 BNC400430-100
BNC400430-500EA 560 EUR
BNC400430-500 BNC400430-100
Anti-CD38 Mouse Monoclonal Antibody (CF640R) [clone: AT1]
Antibodies
CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34 cells. The CD34 CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells).

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR