Anti-KRT76 Mouse Monoclonal Antibody (Biotin) [clone: 34BE12]

Supplier: Biotium
BNCB0446-500 BNCB0446-100
BNCB0446-500EA 560 EUR
BNCB0446-500 BNCB0446-100
Anti-KRT76 Mouse Monoclonal Antibody (Biotin) [clone: 34BE12]
Antibodies
This antibody recognizes CK1, CK5, CK10 and CK14. In normal epithelia, it stains stratified epithelia, myoepithelial cells and basal cells in the prostate gland and bronchi. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody. It stains myoepithelial cells and has been shown to be useful in distinguishing prostate adenocarcinoma from benign prostate. This antibody has also been useful in separating benign from malignant intraductal breast proliferations.

Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: 34BE12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR