Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: 3E2D10 VWF635]
Supplier: Biotium
BNC880646-500 BNC880646-100
BNC880646-500EA 560 EUR
BNC880646-500 BNC880646-100
Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: 3E2D10 VWF635]
Antibodies
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascularMore Product Information