Anti-ACTA2 Rat Monoclonal Antibody (CF680R) [clone: 1A4]

Supplier: Biotium
BNC810665-500 BNC810665-100
BNC810665-500EA 560 EUR
BNC810665-500 BNC810665-100
Anti-ACTA2 Rat Monoclonal Antibody (CF680R) [clone: 1A4]
Antibodies
Actin is a major component of the cytoskeleton and is present in most cell types. This MAb is highly specific to actin from smooth muscles. Its epitope lies in the first four N-terminal amino acids. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: Clone 1A4
Conjugation: CF680R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Baboon, Cat, Chicken, Cow, Dog, Goat, Guinea Pig, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR