Anti-CD68 Mouse Monoclonal Antibody (CF405S) [clone: LAMP4/824]

Supplier: Biotium
BNC040824-500 BNC040824-100
BNC040824-500EA 560 EUR
BNC040824-500 BNC040824-100
Anti-CD68 Mouse Monoclonal Antibody (CF405S) [clone: LAMP4/824]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: LAMP4/824
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR