Anti-CD46 Mouse Monoclonal Antibody (Purified) [clone: 169-1-E4.3]
Supplier: Biotium
BNUB0930-500 BNUB0930-100
BNUB0930-500EA 511 EUR
BNUB0930-500 BNUB0930-100
Anti-CD46 Mouse Monoclonal Antibody (Purified) [clone: 169-1-E4.3]
Antibodies
Recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). Its epitope is located on the four immunogenic short consensus repeat (SCR) domains at N-terminus of the protein. CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virusMore Product Information