Anti-IGHG Mouse Monoclonal Antibody (CF405S) [clone: B33/20]

Supplier: Biotium
BNC040952-500 BNC040952-100
BNC040952-500EA 560 EUR
BNC040952-500 BNC040952-100
Anti-IGHG Mouse Monoclonal Antibody (CF405S) [clone: B33/20]
Antibodies
Recognizes a protein of 75 kDa, identified as γ heavy chain of human immunoglobulins. Its epitope maps in CH2 domain of Fc region of IgG. It reacts with all sub-classes of γ chain of human immunoglobulins. It does not cross-react with α (IgA), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Type: Primary
Antigen: IGHG
Clonality: Monoclonal
Clone: B33/20
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR