Anti-CD84 Mouse Monoclonal Antibody (Purified) [clone: 152-1D5]

Supplier: Biotium
BNUB0985-500 BNUB0985-100
BNUB0985-500EA 511 EUR
BNUB0985-500 BNUB0985-100
Anti-CD84 Mouse Monoclonal Antibody (Purified) [clone: 152-1D5]
Antibodies
Recognizes a protein of 74 kDa, identified as CD84 (Workshop V; Code A085, C057). It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 152-1D5
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR