Anti-CD11B/C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40766SS NB110-40766
NOVUNB110-40766SSEA 280 EUR
NOVUNB110-40766SS NOVUNB110-40766
Anti-CD11B/C Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal CD11b/c Antibody. Tested Applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 5-10 ?g/ml

Type: Primary
Antigen: CD11b/c
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR