Anti-CHGA Mouse Monoclonal Antibody (CF640R) [clone: LK2H10 PHE5 CGA414]

Supplier: Biotium
BNC401223-500 BNC401223-100
BNC401223-500EA 560 EUR
BNC401223-500 BNC401223-100
Anti-CHGA Mouse Monoclonal Antibody (CF640R) [clone: LK2H10 PHE5 CGA414]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: LK2H10 PHE5 CGA414
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR