Cut resistant gloves, HyFlex® 11-651
Supplier: ANSELL HEALTH CARE
11-651/7 11-651/9
ANSE11-651/7EA 0 EUR
ANSE11-651/7 ANSE11-651/9
Cut resistant gloves, HyFlex® 11-651
Gloves
These gloves are made from Polyethylene high tenacity/Lycra® and high performances fibres, using Techcor® Knitting Technology. With black PU coating on palm and fingertips.
  • Seamless knitted glove
  • Fine, very stretchable and comfortable stitches
  • Excellent comfort and dexterity
  • Highest level of cut protection (level 5)
  • Good abrasion resistance
Ideal for handling glass sheets, sharp-edged objects, and metal sheets as well as for cutting dry, painted or galvanised pieces, maintenance and pre-assembling.
Certifications: EN 388:2016 (4X42C); Cat. II