Sodium glucoheptonate dihydrate ≥98.5%

Supplier: ALFA AESAR

J66811.A1 J66811.22 J66811.36
J66811.A1EA 308 EUR
J66811.A1 J66811.22 J66811.36
Sodium glucoheptonate dihydrate ≥98.5%
Sodium glucoheptonate dihydrate

Sodium glucoheptonate is used for cleaning metal, mercerising, paint stripping, and aluminum etching. It is also used to mercerise cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.


  • Freely soluble in water


Caution: Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Formula: C₇H₁₄O₈
MW: 248.16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00013425
CAS Number: 31138-65-5
EINECS: 250-480-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR