tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate 95%

Warning

454620010 454620050
ACRO454620010EA 59.9 EUR
ACRO454620010 ACRO454620050
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate 95%
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Formula: C₁₁H₂₃N₃O₂
Boiling Pt: 198 °C
Melting Pt: 36…41 °C
Storage Temperature: Ambient
MDL Number: MFCD02683049
CAS Number: 192130-34-0

Order Now

Specification Test Results

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR