Bergapten 98%
Supplier: ACROS ORGANICS
|
Danger
|
460660010
460660050
460660250
ACRO460660010EA
67.4
EUR
ACRO460660010
ACRO460660050
ACRO460660250
Bergapten 98%
Bergapten
Formula:
C₁₂H₈O₄ Melting Pt: 190…193 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010272 CAS Number: 484-20-8 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 189 to 193 °C |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...