Theophylline-7-acetic acid 98%

Warning

459250250 459251000
ACRO459250250EA 28.9 EUR
ACRO459250250 ACRO459251000
Theophylline-7-acetic acid 98%
Theophylline-7-acetic acid
Formula: C₉H₁₀N₄O₄
MW: 238.20 g/mol
Melting Pt: 270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022832
CAS Number: 652-37-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 268 to 272 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR