NANOCOLOR® Photometric Tube Tests, COD
Supplier: MACHEREY-NAGEL
985033 985028 963026 985023 985022 985012 985030 985011 985026 985027 985029 963029 985036 985038 985020
MANA985033EA 132 EUR
MANA985033 MANA985028 MANA963026 MANA985023 MANA985022 MANA985012 MANA985030 MANA985011 MANA985026 MANA985027 MANA985029 MANA963029 MANA985036 MANA985038 MANA985020
NANOCOLOR® Photometric Tube Tests, COD
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of the COD (Chemical oxygen demand) in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The tests are particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.