Tin(IV) oxide 15% in water, colloidal dispersion
44592.36
44592.A3
44592.36EA
98.4
EUR
44592.36
44592.A3
Tin(IV) oxide 15% in water, colloidal dispersion
Tin (IV) oxide
CAS No.: 18282-10-5
Molecular Formula: O2Sn
Formula Weight: 150.69
Physical Form: Liquid
Density: 1.153
Odour: Odourless
MDL No.: MFCD00011244
Formula:
O₂Sn Density: 1,153 Storage Temperature: Ambient |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 EINECS: 242-159-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...