Gold ≥99.9% (metals basis), wire, Ø 0.2 mm (0.008 in)
10195.G1
10195.G5
10195.G9
10195.G1EA
199
EUR
10195.G1
10195.G5
10195.G9
Gold ≥99.9% (metals basis), wire, Ø 0.2 mm (0.008 in)
Gold
Purity: 99.9%
CAS No.: 7440-57-5
Physical Form: ≈0.62g/m
Odour: Odourless
MDL No.: MFCD00003436
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...