Tantalum ≥99.9% (metals basis), tubing 3.18 mm (0.125 in) OD, 2.26 mm (0.089 in) ID

Supplier: ALFA AESAR

44923.BW 44923.BS
44923.BWEA 0 EUR
44923.BW 44923.BS
Tantalum ≥99.9% (metals basis), tubing 3.18 mm (0.125 in) OD, 2.26 mm (0.089 in) ID
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Odour: Odourless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR