Polyethylene glycol 6,000
A1387.1000
A1387.5000
A1387.1000EA
69.5
EUR
A1387.1000
A1387.5000
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula:
H(OCH₂CH₂)nOH MW: 6000 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 55…62 °C Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01779614 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...