DL-Lithium lactate 99%, extra pure

413330050 413330250 413331000 413335000
ACRO413330050EA 28.8 EUR
ACRO413330050 ACRO413330250 ACRO413331000 ACRO413335000
DL-Lithium lactate 99%, extra pure
DL-Lithium lactate
Formula: H₃CCH(OH)COOLi
MW: 96.01 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00065399
CAS Number: 867-55-0
EINECS: 212-761-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (10% in water) Clear colorless
pH 6 to 9 (1 % solution)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR