Hydrazine solution 24-26% (by iodometry), Fluka™
Supplier: HONEYWELL
|
Danger
|
15622-1L
15622-6X1L
15622-1LEA
100.51
EUR
15622-1L
15622-6X1L
Hydrazine solution 24-26% (by iodometry), Fluka™
Hydrazine
Beilstein Number: 878137
Formula:
H₄N₂ Density: 1,010 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011417 CAS Number: 302-01-2 EINECS: 206-114-9 UN: 3293 ADR: 6.1,III |
Specification Test Results
Assay (acidimetric) | 24 - 26 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...