Guanidinium hydrochloride
|
Warning
|
A3240.0500
A3240.1000
A3240.0500EA
226
EUR
A3240.0500
A3240.1000
Guanidinium hydrochloride
Guanidinium hydrochloride
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1,354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...