Talc Ø -200 mesh (<75 µm)

Warning

222730025 222735000 222730100
ACRO222730025EA 187 EUR
ACRO222730025 ACRO222735000 ACRO222730100
Talc Ø -200 mesh (<75 µm)
Talc
Formula: Mg₃O₁₁Si₄·1H₂O
MW: 379.27 g/mol
Melting Pt: 800 °C
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Particle size ≤75 µm
Loss on ignition ≤7.0 % (1 g, 1100°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR